missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABBR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528340
This item is not returnable.
View return policy
Description
Recommended positive controls: U87-MG, unboiled Myc-DDK-tagged Gabbr2-transfected 293T. Predicted reactivity: Mouse (97%), Rat (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The multi-pass membrane protein, GABBR2, belongs to the G-protein coupled receptor 3 family and GABA-B receptor subfamily. The GABA-B receptors inhibit neuronal activity through G protein-coupled second-messenger systems, which regulate the release of neurotransmitters, and the activity of ion channels and adenylyl cyclase. This receptor subunit forms an active heterodimeric complex with GABA-B receptor subunit 1, neither of which is effective on its own. Allelic variants of this gene have been associated with nicotine dependence.
Specifications
| GABBR2 | |
| Polyclonal | |
| Unconjugated | |
| GABBR2 | |
| FLJ36928; G protein-coupled receptor 51; GABA BR; GABA-B R2 receptor; GABA-B receptor 2; GABA-B receptor, R2 subunit; GABABR2; GABA-BR2; GABA-B-R2; Gabbr2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) B receptor 2; gamma-aminobutyric acid (GABA) B receptor, 2; gamma-aminobutyric acid B receptor 2; gamma-aminobutyric acid type B receptor subunit 2; gamma-aminobutyric acid type B receptor, subunit 2; gb2; Gm425; Gpr51; GPRC3B; G-protein coupled receptor 51; HG20; HRIHFB2099; MGC119386; MGC119388; MGC119389; ortholog of human G protein-coupled receptor 51 GPR51; RP23-337N2.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83633, 9568 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Western Blot | |
| 0.83 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O75899, O88871 | |
| GABBR2 | |
| Recombinant fragment corresponding to a region within amino acids 163 and 516 of Human GABA B Receptor 2 | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction