missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABBR2 Monoclonal Antibody (S81-2), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA545534
This item is not returnable.
View return policy
Description
1 μg/mL of MA5-45534 was sufficient for detection of GABA(B)R2 in 20 μg of rat brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.∣Detects approximately 105kDa. No cross-reactivity against GABA(B)R1.
The multi-pass membrane protein, GABBR2, belongs to the G-protein coupled receptor 3 family and GABA-B receptor subfamily. The GABA-B receptors inhibit neuronal activity through G protein-coupled second-messenger systems, which regulate the release of neurotransmitters, and the activity of ion channels and adenylyl cyclase. This receptor subunit forms an active heterodimeric complex with GABA-B receptor subunit 1, neither of which is effective on its own. Allelic variants of this gene have been associated with nicotine dependence.Specifications
GABBR2 | |
Monoclonal | |
1 mg/mL | |
9.1mM sodium bicarbonate, 9.44mM PBS with 123mM ethanolamine, 581mM DMSO and no preservative; pH 7.4 | |
O75899, O88871, Q80T41 | |
GABBR2 | |
Fusion protein amino acids 861-912 of rat GABA(B)R2 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
S81-2 | |
FITC | |
GABBR2 | |
FLJ36928; G protein-coupled receptor 51; GABA BR; GABA-B R2 receptor; GABA-B receptor 2; GABA-B receptor, R2 subunit; GABABR2; GABA-BR2; GABA-B-R2; Gabbr2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) B receptor 2; gamma-aminobutyric acid (GABA) B receptor, 2; gamma-aminobutyric acid B receptor 2; gamma-aminobutyric acid type B receptor subunit 2; gamma-aminobutyric acid type B receptor, subunit 2; gb2; Gm425; Gpr51; GPRC3B; G-protein coupled receptor 51; HG20; HRIHFB2099; MGC119386; MGC119388; MGC119389; ortholog of human G protein-coupled receptor 51 GPR51; RP23-337N2.1 | |
Mouse | |
Protein G | |
RUO | |
242425, 83633, 9568 | |
4° C, store in dark | |
IgG1 |