missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABBR2 Monoclonal Antibody (5G7F7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA538528
This item is not returnable.
View return policy
Description
This antibody has been tested in indirect ELISA.
The multi-pass membrane protein, GABBR2, belongs to the G-protein coupled receptor 3 family and GABA-B receptor subfamily. The GABA-B receptors inhibit neuronal activity through G protein-coupled second-messenger systems, which regulate the release of neurotransmitters, and the activity of ion channels and adenylyl cyclase. This receptor subunit forms an active heterodimeric complex with GABA-B receptor subunit 1, neither of which is effective on its own. Allelic variants of this gene have been associated with nicotine dependence.
Specifications
| GABBR2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O75899 | |
| GABBR2 | |
| Purified recombinant fragment of human GABBR2 (aa: 319-483) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 5G7F7 | |
| Unconjugated | |
| GABBR2 | |
| FLJ36928; G protein-coupled receptor 51; GABA BR; GABA-B R2 receptor; GABA-B receptor 2; GABA-B receptor, R2 subunit; GABABR2; GABA-BR2; GABA-B-R2; Gabbr2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) B receptor 2; gamma-aminobutyric acid (GABA) B receptor, 2; gamma-aminobutyric acid B receptor 2; gamma-aminobutyric acid type B receptor subunit 2; gamma-aminobutyric acid type B receptor, subunit 2; gb2; Gm425; Gpr51; GPRC3B; G-protein coupled receptor 51; HG20; HRIHFB2099; MGC119386; MGC119388; MGC119389; ortholog of human G protein-coupled receptor 51 GPR51; RP23-337N2.1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 9568 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction