missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gabbr1, Mouse anti-Rat, PerCP, Clone: S93A-49, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB18285.100ug
This item is not returnable.
View return policy
Specifications
| Gabbr1 | |
| Monoclonal | |
| PerCP | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
| - | |
| Mouse | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| S93A-49 | |
| Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| gamma-aminobutyric acid (GABA) B receptor 1 | |
| Gabbr1 | |
| Recombinant protein corresponding to amino acids 873-977 at C-terminus of rat Gabbr1. | |
| RUO | |
| 81657 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction