missing translation for 'onlineSavingsMsg'
Learn More

Gabazine, MedChemExpress

MedChemExpress Gabazine is a selective and competitive antagonist of GABAA receptor, with an IC50 of ∼0.2 μM for GABA receptor.

£72.60 - £709.06

Specifications

Chemical Name or Material Gabazine
CAS 104104-50-9
Color White
Purity Grade Notes Research
Molecular Formula C15H18BrN3O3
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16473858
Encompass_Preferred
View Documents
Medchem Express
HY-103533
10 mg
£107.69
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16446587
Encompass_Preferred
View Documents
Medchem Express
HY-103533
5 mg
£72.60
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16404896
Encompass_Preferred
View Documents
Medchem Express
HY-103533
100 mg
£709.06
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16400237
Encompass_Preferred
View Documents
Medchem Express
HY-103533
50 mg
£396.88
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gabazine is a selective and competitive antagonist of GABAA receptor, with an IC50 of ∼0.2 μM for GABA receptor.
Specifications

Specifications

Gabazine
White
C15H18BrN3O3
H2O : 100 mg/mL (271.57 mM; Need ultrasonic) ∣DMSO : ≥ 75 mg/mL (203.68 mM)
368.23
98.0%
Powder
104104-50-9
Research
SR95531
O=C(O)CCCN1C(C=CC(C2=CC=C(OC)C=C2)=N1)=N.[H]Br
368.23
Research
Neuroscience-Neuromodulation
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.