missing translation for 'onlineSavingsMsg'
Learn More

Gabazine, MedChemExpress

Product Code. 16400237
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16400237

Brand: Medchem Express HY103533

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gabazine is a selective and competitive antagonist of GABAA receptor, with an IC50 of ∼0.2 μM for GABA receptor.

Gabazine is a selective and competitive antagonist of GABAA receptor, with an IC50 of ∼0.2 μM for GABA receptor.

Specifications

Chemical Name or Material Gabazine
CAS 104104-50-9
Color White
Purity Grade Notes Research
Molecular Formula C15H18BrN3O3
Quantity 50 mg
Synonym SR95531
Solubility Information H2O : 100 mg/mL (271.57 mM; Need ultrasonic) ∣DMSO : ≥ 75 mg/mL (203.68 mM)
SMILES O=C(O)CCCN1C(C=CC(C2=CC=C(OC)C=C2)=N1)=N.[H]Br
Molecular Weight (g/mol) 368.23
Formula Weight 368.23
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.