missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABARAPL1 Recombinant Rabbit Monoclonal Antibody (5P10L6)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558105
This item is not returnable.
View return policy
Description
Immunogen sequence: MKFQYKEDHP FEYRKKEGEK IRKKYPDRVP VIVEKAPKAR VPDLDKRKYL VPSDLTVGQF YFLIRKRIHL RPEDALFFFV NNTIPPTSAT MGQLYEDNHE.
GABARAPL1 increases cell-surface expression of kappa-type opioid receptor through facilitating anterograde intracellular trafficking of the receptor.
Specifications
| GABARAPL1 | |
| Recombinant Monoclonal | |
| 1.92 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q0VGK0, Q8R3R8, Q9H0R8 | |
| GABARAPL1 | |
| Synthetic peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 5P10L6 | |
| Unconjugated | |
| GABARAPL1 | |
| 3110025G09Rik; 9130422N19Rik; AI196471; Apg8l; APG8LIKE; APG8-LIKE; ATG8; ATG8B; Atg8l; Early estrogen-regulated protein; GABA type A receptor associated protein like 1; GABA(A) receptor-associated protein like 1; GABA(A) receptor-associated protein-like 1; GABA(A)-RAP; GABARAPL1; GABARAPL1-a; gamma-aminobutyric acid (GABA(A)) receptor-associated protein-like 1; gamma-aminobutyric acid (GABA) A receptor-associated protein-like 1; gamma-aminobutyric acid receptor-associated protein-like 1; gamma-aminobutyric acid-receptor-associated protein; Gec1; GEC-1; GECI; Glandular epithelial cell protein 1; MDBK gamma-aminobutyric acid-receptor-associated protein; MNCb-0091 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23710, 57436, 689161 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction