missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GABARAPL1 Recombinant Rabbit Monoclonal Antibody (5P10L6)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558106

 View more versions of this product

Product Code. 30282855

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MKFQYKEDHP FEYRKKEGEK IRKKYPDRVP VIVEKAPKAR VPDLDKRKYL VPSDLTVGQF YFLIRKRIHL RPEDALFFFV NNTIPPTSAT MGQLYEDNHE.

GABARAPL1 increases cell-surface expression of kappa-type opioid receptor through facilitating anterograde intracellular trafficking of the receptor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GABARAPL1
Recombinant Monoclonal
1.92 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q0VGK0, Q8R3R8, Q9H0R8
GABARAPL1
Synthetic peptide
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot
5P10L6
Unconjugated
GABARAPL1
3110025G09Rik; 9130422N19Rik; AI196471; Apg8l; APG8LIKE; APG8-LIKE; ATG8; ATG8B; Atg8l; Early estrogen-regulated protein; GABA type A receptor associated protein like 1; GABA(A) receptor-associated protein like 1; GABA(A) receptor-associated protein-like 1; GABA(A)-RAP; GABARAPL1; GABARAPL1-a; gamma-aminobutyric acid (GABA(A)) receptor-associated protein-like 1; gamma-aminobutyric acid (GABA) A receptor-associated protein-like 1; gamma-aminobutyric acid receptor-associated protein-like 1; gamma-aminobutyric acid-receptor-associated protein; Gec1; GEC-1; GECI; Glandular epithelial cell protein 1; MDBK gamma-aminobutyric acid-receptor-associated protein; MNCb-0091
Rabbit
Protein A
RUO
23710, 57436, 689161
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.