missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABARAP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18723-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody detected endogenous GABARAP protein in lysates from various tissues. It may cross-react with GABARAPL1 due to the high identity of amino acid sequence.
Gamma-aminobutyric acid (GABA) is the main inhibitory transmitter by increasing a Cl-conductance that inhibits neuronal firing in the central nervous system. It has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. GABARAP (GABAA receptor-associated protein) links GABAA receptors to the cytoskeleton and may play a role in intracellular transport of GABAA receptors and its interaction with the cytoskeleton. GABARAP belongs to the MAP1 or ATG8 like family and recent studies show that MAPK15/ERK8 is acting through interaction with ATG8 family proteins to regulate autophagy.Specifications
| GABARAP | |
| Polyclonal | |
| Unconjugated | |
| GABARAP | |
| FLC3B, GABARAP, MM46 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11337, 56486, 58974 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95166, P60517, Q9DCD6 | |
| Gabarap | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction