missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABARAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578366
This item is not returnable.
View return policy
Description
Positive Control: 293T, U87-MG, SK-N-SH, IMR32, SK-N-AS Predicted Reactivity: Mouse (100%), Rat (100%), Pig (100%), Rabbit (100%), Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Gamma-aminobutyric acid (GABA) is the main inhibitory transmitter by increasing a Cl-conductance that inhibits neuronal firing in the central nervous system. It has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. GABARAP (GABAA receptor-associated protein) links GABAA receptors to the cytoskeleton and may play a role in intracellular transport of GABAA receptors and its interaction with the cytoskeleton. GABARAP belongs to the MAP1 or ATG8 like family and recent studies show that MAPK15/ERK8 is acting through interaction with ATG8 family proteins to regulate autophagy.
Specifications
| GABARAP | |
| Polyclonal | |
| Unconjugated | |
| GABARAP | |
| ATG8A; FLC3B; FLJ25768; GABA type A receptor-associated protein; GABA(A) receptor-associated protein; GABA-A receptor-associated protein; Gabarap; GABARAP a; GABARAP-a; gamma-aminobutyric acid receptor associated protein; gamma-aminobutyric acid receptor-associated protein; gamma-aminobutyric acid reseptor associated protein; HT004; hypothetical protein; MGC120154; MGC120155; MM46 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11337 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 2.48 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O95166 | |
| GABARAP | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human GABARAP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction