missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABARAP Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71771B
This item is not returnable.
View return policy
Description
GABARAP Polyclonal specifically detects GABARAP in Human, Mouse, Drosophila samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| GABARAP | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| O95166 | |
| GABARAP | |
| A synthetic peptide made to a C-terminal region of the human GABARAP protein (within residues 50-117). [Swiss-Prot O95166] | |
| 0.1 mL | |
| Autophagy, Golgi Apparatus Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 11337 | |
| Human, Mouse, Drosophila | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| ATG8A, FLC3B, FLJ25768, GABA(A) receptor-associated proteinMGC120154, gamma-aminobutyric acid receptor-associated protein, MM46MGC120155 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Predicted to react with rat based on 100% sequence homology. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction