missing translation for 'onlineSavingsMsg'
Learn More

Gabapentin, TRC

Product Code. 30389213
Change view
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389213 10 mg -
30390649 50 mg -
30399030 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389213 Supplier Toronto Research Chemicals Supplier No. TRCG11725010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60142-96-3
Molecular Formula C9 H17 N O2
Molecular Weight (g/mol) 171.24
Synonym 1-(Aminomethyl)-cyclohexaneacetic acid, CI 945, GOE 2450, GOE 3450, Gabapen, Gabapentin, Go 3450, Neurontin
IUPAC Name 2-[1-(aminomethyl)cyclohexyl]acetic acid
SMILES NCC1(CC(=O)O)CCCCC1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H17NO2/c10-7-9(6-8(11)12)4-2-1-3-5-9/h1-7,10H2,(H,11,12)
Formula Weight 171.1259
Percent Purity >95%
Chemical Name or Material Gabapentin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.