missing translation for 'onlineSavingsMsg'
Learn More

Gabapentin Related Compound E, TRC

Product Code. 17930485
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17930485 25 mg 25mg
30136520 100 mg 100mg
17974625 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17930485 Supplier Toronto Research Chemicals Supplier No. TRCG11729025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67950-95-2
Molecular Formula C9 H14 O4
Molecular Weight (g/mol) 186.21
Synonym 1-(Carboxymethyl)cyclohexanecarboxylic Acid, Gabapentin Imp. E (EP)
IUPAC Name 1-(carboxymethyl)cyclohexane-1-carboxylic acid
SMILES OC(=O)CC1(CCCCC1)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C9H14O4/c10-7(11)6-9(8(12)13)4-2-1-3-5-9/h1-6H2,(H,10,11)(H,12,13)
Formula Weight 186.0892
Percent Purity >95%
Chemical Name or Material 1-(Carboxymethyl)cyclohexanecarboxylic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.