missing translation for 'onlineSavingsMsg'
Learn More

Gabapentin Related Compound D, TRC

Product Code. 30392931
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392931 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392931 Supplier Toronto Research Chemicals Supplier No. TRCG1172852.5MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material [1-[(3-Oxo-2-azaspiro[4.5]dec-2-yl)methyl]cyclohexyl]acetic Acid
CAS 1076198-17-8
Molecular Formula C18 H29 N O3
Quantity 2.5 mg
Synonym Cyclohexaneacetic acid, 1-[(3-oxo-2-azaspiro[4.5]dec-2-yl)methyl]-, [1-[(3-oxo-2-azaspiro[4.5]dec-2-yl)methyl]cyclohexyl]acetic acid, Gabapentin Imp. D (EP), Gabapentin RC D (USP), [1-[(3-Oxo-2-azaspiro[4.5]dec-2-yl)methyl]cyclohexyl]acetic Acid
InChI Formula InChI=1S/C18H29NO3/c20-15-11-17(7-3-1-4-8-17)13-19(15)14-18(12-16(21)22)9-5-2-6-10-18/h1-14H2,(H,21,22)
SMILES OC(=O)CC1(CN2CC3(CCCCC3)CC2=O)CCCCC1
IUPAC Name 2-[1-[(3-oxo-2-azaspiro[4.5]decan-2-yl)methyl]cyclohexyl]acetic acid
Molecular Weight (g/mol) 307.43
Formula Weight 307.2147
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.