missing translation for 'onlineSavingsMsg'
Learn More

Gabapentin, MedChemExpress

Product Code. 16478278
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478278 10 mg 10mg
16468006 100 mg 100mg
16454167 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478278 Supplier Medchem Express Supplier No. HYA0057

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gabapentin (Neurontin) is a pharmaceutical drug, specifically a GABA analog.

Gabapentin (Neurontin) is a pharmaceutical drug, specifically a GABA analog.

Specifications

Chemical Name or Material Gabapentin
Physical Form Solid
CAS 60142-96-3
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C9H17NO2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information H2O : 50 mg/mL (291.99 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (5.84 mM; Need ultrasonic)
SMILES O=C(O)CC1(CN)CCCCC1
Molecular Weight (g/mol) 171.24
Formula Weight 171.24
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.