missing translation for 'onlineSavingsMsg'
Learn More

Gabapentin Ethyl Ester Hydrochloride, TRC

Product Code. 30394613
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394613 250 mg -
30396123 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394613 Supplier Toronto Research Chemicals Supplier No. TRCG117255250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60175-04-4
Molecular Formula C11 H21 N O2 . Cl H
Molecular Weight (g/mol) 235.75
Synonym 1-(Aminomethyl)-cyclohexaneacetic acid ethyl ester hydrochloride (1:1), Cyclohexaneacetic acid, 1-(aminomethyl)-, ethyl ester, hydrochloride (9CI)
IUPAC Name ethyl 2-[1-(aminomethyl)cyclohexyl]acetate;hydrochloride
SMILES Cl.CCOC(=O)CC1(CN)CCCCC1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H21NO2.ClH/c1-2-14-10(13)8-11(9-12)6-4-3-5-7-11;/h2-9,12H2,1H3;1H
Formula Weight 235.1339
Chemical Name or Material Gabapentin Ethyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.