missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gabapentin Ethyl Ester Hydrochloride, Mikromol™
Description
Gabapentin Ethyl Ester Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Gabapentin Ethyl Ester Hydrochloride |
| CAS | 60175-04-4 |
| Analyte or Component Names | Gabapentin Ethyl Ester Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H21 N O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-(Aminomethyl)-cyclohexaneacetic acid ethyl ester hydrochloride (1:1), Cyclohexaneacetic acid, 1-(aminomethyl)-, ethyl ester, hydrochloride (9CI) |
| InChI Formula | InChI=1S/C11H21NO2.ClH/c1-2-14-10(13)8-11(9-12)6-4-3-5-7-11;/h2-9,12H2,1H3;1H |
| SMILES | Cl.CCOC(=O)CC1(CN)CCCCC1 |
| IUPAC Name | ethyl 2-[1-(aminomethyl)cyclohexyl]acetate;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?