missing translation for 'onlineSavingsMsg'
Learn More

Gabapentin enacarbil, MedChemExpress

Product Code. 16447008
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16447008

Brand: Medchem Express HY16216

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gabapentin enacarbil (XP-13512) is a prodrug for the anticonvulsant and analgesic drug gabapentin.

Gabapentin enacarbil (XP-13512) is a prodrug for the anticonvulsant and analgesic drug gabapentin.

Specifications

Chemical Name or Material Gabapentin enacarbil
Physical Form Viscous Liquid
CAS 478296-72-9
Color Light Brown
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H27NO6
For Use With (Application) Neuroscience-Neuromodulation
Synonym XP-13512
Solubility Information Ethanol : 100 mg/mL (303.59 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (303.59 mM) ∣H2O : 0.67 mg/mL (2.03 mM; Need ultrasonic)
SMILES O=C(O)CC1(CNC(OC(OC(C(C)C)=O)C)=O)CCCCC1
Molecular Weight (g/mol) 329.39
Formula Weight 329.39
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.