missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gabapentin-d4 (100 μg/mL in Methanol), TRC
Specifications
Specifications
| CAS | 1185039-20-6 |
| Molecular Formula | C9 D4 H13 N O2 |
| Quantity | 5 x 1 mL |
| Synonym | Gabapentin-d4, Gabapentin D4 (aminomethyl D2, acetic acid D2), Cyclohexaneacetic-α,α-d2 acid, 1-(aminomethyl-d2)- (ACI), 1-(Aminomethyl-d2)cyclohexaneacetic-α,α-d2 acid (ACI), 2-[1-(Amino-dideuteriomethyl)cyclohexyl]-2,2-dideuterioacetic acid |
| InChI Formula | InChI=1S/C9H17NO2/c10-7-9(6-8(11)12)4-2-1-3-5-9/h1-7,10H2,(H,11,12)/i6D2,7D2 |
| SMILES | [2H]C([2H])(N)C1(CCCCC1)C([2H])([2H])C(=O)O |
| IUPAC Name | 2-[1-[amino(dideuterio)methyl]cyclohexyl]-2,2-dideuterioacetic acid |
| Molecular Weight (g/mol) | 175.261 |
| Formula Weight | 175.151 |
| Concentration or Composition (by Analyte or Components) | Gabapentin-d4 100 μg/mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?