missing translation for 'onlineSavingsMsg'
Learn More

Gabapentin 100 μg/mL in Acetonitrile:Methanol, Dr. Ehrenstorfer

Product Code. 30311031
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30311031 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311031 Supplier Dr. Ehrenstorfer Supplier No. DREA13991500MC100

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gabapentin
CAS 60142-96-3
Analyte or Component Names Gabapentin
Shipping Condition Room Temperature
Molecular Formula C9 H17 N O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 1-(Aminomethyl)-cyclohexaneacetic acid, CI 945, GOE 2450, GOE 3450, Gabapen, Gabapentin, Go 3450, Neurontin
InChI Formula InChI=1S/C9H17NO2/c10-7-9(6-8(11)12)4-2-1-3-5-9/h1-7,10H2,(H,11,12)
SMILES NCC1(CC(=O)O)CCCCC1
IUPAC Name 2-[1-(aminomethyl)cyclohexyl]acetic acid
Solvent or Matrix Acetonitrile/Methanol
Molecular Weight (g/mol) 171.24
Quantity 1 mL
Formula Weight 171.1259 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.