missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-B R1, p Ser923 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-61102
This item is not returnable.
View return policy
Description
GABA-B R1 Polyclonal specifically detects GABA-B R1 in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| GABA-B R1 (p Ser923) | |
| Polyclonal | |
| Western Blot 1:1000 | |
| Q9Z0U4 | |
| GABBR1 | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser923 conjugated to KLH. Accession # Q9Z0U4 | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Specific for approx. 102k GABAB R1 phosphorylated at Ser. Immunolabeling of the GABA923B R1 band is completely blocked by gamma-phosphatase treatment. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| FLJ92613, GABABR1, GABBR1-3, gamma-aminobutyric acid (GABA) B receptor, 1, gamma-aminobutyric acid type B receptor subunit 1, GB1, GPRC3Asubunit 1c, hGB1a, seven transmembrane helix receptor | |
| Rabbit | |
| 102 kDa | |
| 0.1 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific | |
| 2550 | |
| Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction