missing translation for 'onlineSavingsMsg'
Learn More

GABA-B R1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£292.00 - £488.00

Specifications

Antigen GABA-B R1
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18408911
View Documents
Novus Biologicals
NBP2-14033
0.1 mL
£488.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18430482
View Documents
Novus Biologicals
NBP2-14033-25ul
25 μL
£292.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GABA-B R1 Polyclonal antibody specifically detects GABA-B R1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

GABA-B R1
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
PBS (pH 7.2) and 40% Glycerol
2550
IgG
Immunogen affinity purified
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Purified
RUO
Human
FLJ92613, GABABR1, GABBR1-3, gamma-aminobutyric acid (GABA) B receptor, 1, gamma-aminobutyric acid type B receptor subunit 1, GB1, GPRC3Asubunit 1c, hGB1a, seven transmembrane helix receptor
This antibody was developed against a recombinant protein corresponding to the amino acids: YKERLFGKKYVWFLIGWYADNWFKIYDPSINCTVDEMTEAVEGHITTEIVMLNPANTRSISNMTSQEFVEKLTKRLKR
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.