missing translation for 'onlineSavingsMsg'
Learn More

GABA-B R1 Antibody [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30738164 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30738164 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738164 Supplier Novus Biologicals Supplier No. AF7000JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

GABA-B R1 Polyclonal antibody specifically detects GABA-B R1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABA-B R1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias FLJ92613, GABABR1, GABBR1-3, gamma-aminobutyric acid (GABA) B receptor, 1, gamma-aminobutyric acid type B receptor subunit 1, GB1, GPRC3Asubunit 1c, hGB1a, seven transmembrane helix receptor
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant rat GABAB R1, Gly17-Leu586, Accession # Q9Z0U4
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2550
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.