missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-B R1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-160CL1
This item is not returnable.
View return policy
Description
GABA-B R1 Polyclonal antibody specifically detects GABA-B R1 in Human samples. It is validated for Western Blot
Specifications
| GABA-B R1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| FLJ92613, GABABR1, GABBR1-3, gamma-aminobutyric acid (GABA) B receptor, 1, gamma-aminobutyric acid type B receptor subunit 1, GB1, GPRC3Asubunit 1c, hGB1a, seven transmembrane helix receptor | |
| Synthetic peptide corresponding to residues 33-54 [PHLPRPHPRVPPHPSSERRAVY] of the GABABR1 subunit. | |
| 0.1 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific | |
| 2550 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction