missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-AR alpha 2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£171.00 - £385.00
Specifications
| Antigen | GABA-AR alpha 2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18371846
|
Novus Biologicals
NBP3-02985-20UL |
20 μL |
£171.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18371027
|
Novus Biologicals
NBP3-02985-100UL |
100 μL |
£385.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GABA-AR alpha 2 Polyclonal specifically detects GABA-AR alpha 2 in Human, Mouse samples. It is validated for Western Blot.Specifications
| GABA-AR alpha 2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| GABA(A) receptor subunit alpha-2, gamma-aminobutyric acid (GABA) A receptor, alpha 2, gamma-aminobutyric acid receptor subunit alpha-2 | |
| A synthetic peptide corresponding to a sequence within amino acids 300-400 of human GABA-AR alpha 2 (NP_000798.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| PBS with 50% glycerol, pH7.3. | |
| 2555 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title