missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-AR alpha 2 Antibody (S399-19), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59325JF549
This item is not returnable.
View return policy
Description
GABA-AR alpha 2 Monoclonal antibody specifically detects GABA-AR alpha 2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| GABA-AR alpha 2 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| GABA(A) receptor subunit alpha-2, gamma-aminobutyric acid (GABA) A receptor, alpha 2, gamma-aminobutyric acid receptor subunit alpha-2 | |
| Fusion protein amino acids 350-385 (Cytoplasmic C-terminus) of rat GABA-A Receptor Alpha2 | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 2555 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunofluorescence | |
| S399-19 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction