missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-AR alpha 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-36560
This item is not returnable.
View return policy
Description
GABA-AR alpha 2 Polyclonal specifically detects GABA-AR alpha 2 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.
Specifications
| GABA-AR alpha 2 | |
| Polyclonal | |
| Western Blot 1:1000, Immunohistochemistry | |
| GABA(A) receptor subunit alpha-2, gamma-aminobutyric acid (GABA) A receptor, alpha 2, gamma-aminobutyric acid receptor subunit alpha-2 | |
| Rabbit | |
| 51 kDa | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 2555 | |
| Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| GABRA2 | |
| Fusion protein from the cytoplasmic loop of the alpha 2 subunit. Accession # P23576 | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Specific for the ∼51k alpha2-subunit of the GABAA receptor in Western blots. Labeling is absent in alpha2-subunit knockout animals. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction