missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-AR alpha 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB300-207
This item is not returnable.
View return policy
Description
GABA-AR alpha 1 Polyclonal specifically detects GABA-AR alpha 1 in Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation, Knockout Validated.
Specifications
| GABA-AR alpha 1 | |
| Polyclonal | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| ECA4, EJM, EJM5GABA(A) receptor subunit alpha-1, gamma-aminobutyric acid (GABA) A receptor, alpha 1, gamma-aminobutyric acid receptor subunit alpha-1 | |
| Rabbit | |
| 51 kDa | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 2554 | |
| Mouse, Rat | |
| IgG |
| Western Blot, Immunoprecipitation, Immunoassay | |
| 0.15 mg/mL | |
| Western Blot 1:1000, Immunoprecipitation, Knockout Validated | |
| P62813 | |
| GABRA1 | |
| Fusion protein from the cytoplasmic loop of the alpha 1 subunit. Accession # P62813 | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Specific for the ∼51k alpha1-subunit of the GABAA receptor in Western blots. Labeling is absent in alpha1-subunit knockout animals. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction