missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-A R rho 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03696-100UL
This item is not returnable.
View return policy
Description
GABA-A R rho 1 Polyclonal specifically detects GABA-A R rho 1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| GABA-A R rho 1 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2569 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| GABA(C) receptor, gamma-aminobutyric acid (GABA) A receptor, rho-1, gamma-aminobutyric acid (GABA) receptor, rho 1, gamma-aminobutyric acid receptor subunit rho-1, rho 1) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 365-465 of human GABA-A R rho 1 (NP_002033.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction