missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-A R gamma 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Description
GABA-A R gamma 1 Polyclonal antibody specifically detects GABA-A R gamma 1 in Human samples. It is validated for Immunofluorescence
Specifications
Specifications
| Antigen | GABA-A R gamma 1 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
| Formulation | PBS, pH 7.2, 40% glycerol |
| Gene Alias | DKFZp686H2042, GABA(A) receptor subunit gamma-1, gamma-1 polypeptide, gamma-aminobutyric acid (GABA) A receptor, gamma 1, gamma-aminobutyric acid receptor subunit gamma-1, MGC33838 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: NKASMTPGLHPGSTLIPMNNISVPQEDDYGYQCLEGKDC |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?