missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-A R delta Antibody (N151/3), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47616F
This item is not returnable.
View return policy
Description
GABA-A R delta Monoclonal specifically detects GABA-A R delta in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| GABA-A R delta | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| EIG10, EJM7, GABA(A) receptor subunit delta, GABA-A receptor, delta polypeptide, gamma-aminobutyric acid (GABA) A receptor, delta, gamma-aminobutyric acid receptor subunit delta, GEFSP5, MGC45284 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Detects approx 55kDa. | |
| Store at 4C in the dark. | |
| IgG2a |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S151-3 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| P18506 | |
| GABRD | |
| Synthetic peptide amino acids 15-34 of rat GABA-A-R-Delta | |
| 0.1 mL | |
| 2563 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction