missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-A R delta Antibody (N151/3), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47616AF647
This item is not returnable.
View return policy
Description
GABA-A R delta Monoclonal specifically detects GABA-A R delta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| GABA-A R delta | |
| Monoclonal | |
| Alexa Fluor 647 | |
| EIG10, EJM7, GABA(A) receptor subunit delta, GABA-A receptor, delta polypeptide, gamma-aminobutyric acid (GABA) A receptor, delta, gamma-aminobutyric acid receptor subunit delta, GEFSP5, MGC45284 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 2563 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S151-3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| GABRD | |
| Synthetic peptide amino acids 15-34 of rat GABA-A-R-Delta | |
| 0.1 mL | |
| Primary | |
| Detects approx 55kDa. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction