missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-A R beta 3 Antibody (N87/25), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47613IR
This item is not returnable.
View return policy
Description
GABA-A R beta 3 Monoclonal specifically detects GABA-A R beta 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| GABA-A R beta 3 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ECA5, GABA(A) receptor beta-3 subunit, GABA(A) receptor subunit beta-3, GABAA receptor beta-3 subunit, GABA-alpha receptor beta-2 subunit, gamma-aminobutyric acid (GABA) A receptor, beta 3, gamma-aminobutyric acid receptor subunit beta-3, MGC9051 | |
| Mouse | |
| Protein G purified | |
| Cellular Markers, Neuronal Cell Markers, Neurotransmission | |
| 2562 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| S87-25 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| P63080 | |
| GABRB3 | |
| Fusion protein amino acids 370-433 of mouse GABA-A-R-Beta 3 | |
| 0.1 mL | |
| Primary | |
| Detects approx 55kDa. No cross-reactivity against GABA-A-R-Beta 2 or -Beta1. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction