missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-A R beta 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB300-198-0.025ml
This item is not returnable.
View return policy
Description
GABA-A R beta 2 Polyclonal specifically detects GABA-A R beta 2 in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| GABA-A R beta 2 | |
| Polyclonal | |
| Western Blot 1:1000 | |
| P63138 | |
| GABRB2 | |
| Fusion protein from the cytoplasmic loop of the beta 2 subunit. Accession # P63138 | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Specific for the ∼55k beta 2-subunit of the GABAA receptor in Western blots. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| GABA(A) receptor subunit beta-2, gamma-aminobutyric acid (GABA) A receptor, beta 2, gamma-aminobutyric acid A receptor beta 2, gamma-aminobutyric acid receptor subunit beta-2, MGC119386, MGC119388, MGC119389 | |
| Rabbit | |
| 55 kDa | |
| 0.025 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 2561 | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction