missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAB4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560782
This item is not returnable.
View return policy
Description
Immunogen sequence: ESTGFLGNIS SASHGLCSSP AEPSCSHQHL PQEQEPTSEP PVSHCVPPTW PIPAPPGCLR SHQHASQRAE Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 44%.
Specifications
| GAB4 | |
| Polyclonal | |
| Unconjugated | |
| GAB4 | |
| GAB2-like; GAB4; GRB2 associated binding protein family member 4; GRB2-associated binder 2-like; GRB2-associated binder 2-like protein; GRB2-associated binder 4; GRB2-associated binding protein family, member 4; GRB2-associated-binding protein 2-like; GRB2-associated-binding protein 4; growth factor receptor bound protein 2-associated protein 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 128954 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q2WGN9 | |
| GAB4 | |
| Recombinant protein corresponding to Human GAB4. Recombinant protein control fragment (Product #RP-95738). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction