missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAB3 Monoclonal Antibody (720306)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561767
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
GAB3 belongs to the GAB family. GAB3 interacts with PIK3R/p85, SHP2 and GRAP2/MONA. It may interact with Grb2.
Specifications
| GAB3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8WWW8 | |
| GAB3 | |
| E. coli-derived recombinant human GAB3, Leu121-Asn263. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 720306 | |
| Unconjugated | |
| GAB3 | |
| 5930433H21Rik; DOS/Gab family member 3; GAB3; Gab3 scaffolding protein; GRB2 associated binding protein 3; GRB2-associated binder 3; GRB2-associated binding protein 3; GRB2-associated-binding protein 3; Growth factor receptor bound protein 2-associated protein 3 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 139716 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction