Learn More
GAB2 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | GAB2 |
|---|---|
| Concentration | 400 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17231446
|
Proteintech
29272-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17221446
|
Proteintech
29272-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Growth factor receptor-bound protein 2 (GAB2), an adaptor protein, belongs to the Gab family and is involved in various biologic processes. Gab2 mainly mediates phosphatidylinositol 3-kinase (PI3K) and extracellular signal-regulated kinase (ERK) signaling pathways. Furthermore, Gab2 governs tumorigenic signaling and is a novel potential therapeutic target in leukemia, breast cancer, ovarian cancer, and melanoma. Gab2 is the major 98-kDa phosphoprotein associated with SHP-2, PI 3-kinase, and Grb2 in normal human lymphocytes. (PMID: 32269703, PMID: 28842424, PMID: 28420432, PMID: 10849428)
Specifications
| GAB2 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Rabbit | |
| 9846 |
| 400 μg/mL | |
| Polyclonal | |
| Liquid | |
| Human | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.