missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAB1 Antibody (1A7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£199.00 - £439.00
Specifications
| Antigen | GAB1 |
|---|---|
| Clone | 1A7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18679035
|
Novus Biologicals
NBP2-37742-0.025ml |
0.025 mL |
£199.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18770244
|
Novus Biologicals
NBP2-37742 |
0.1 mL |
£439.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GAB1 Monoclonal specifically detects GAB1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| GAB1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| GRB2-associated binder 1, GRB2-associated binding protein 1, GRB2-associated-binding protein 1, Growth factor receptor bound protein 2-associated protein 1 | |
| GAB1 | |
| IgG1 | |
| Affinity Purified |
| 1A7 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Apoptosis, Signal Transduction, Tyrosine Kinases | |
| PBS with 0.05% Sodium Azide | |
| 2549 | |
| Purified recombinant fragment of human GAB1 expressed in E. Coli. | |
| Primary | |
| 76.6 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title