missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ G6PDi 1
GREENER_CHOICE

Product Code. 30000024
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000024 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000024 Supplier Tocris Bioscience™ Supplier No. 7561/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, reversible glucose-6-phosphate dehydrogenase (G6PD) inhibitor

G6PDi 1 is a potent, reversible, nonsteroidal inhibitor of glucose-6-phosphate dehydrogenase (G6PD) (IC50 = 70 nM. The compound dose-dependently and reversibly decreases 6-phosphogluconolactone (6-PG) levels in HCT116 and HepG2 cell lines in vitro. In activated T cells, G6PDi-1 inhibits flux through the pentose phosphate pathway and decreases NADPH generation. It also inhibits production of cytokines by activated T cells and oxidative burst in neutrophils.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-((5-Oxo-6,7,8,9-tetrahydro-5H-cyclohepta[d]pyrimidin-2-yl)amino)thiophene-2-carbonitrile
CAS 2457232-14-1
Quantity 10 mg
Target Glucose 6 Phosphate Dehydrogenase
Molecular Formula C14H12N4OS
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.