missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ G6PD Recombinant Rabbit Monoclonal Antibody (K01_1R70)

Product Code. 30825404
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825404 100 μL
30825119 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825404 Supplier Invitrogen™ Supplier No. MA563471

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

G6PD Recombinant Monoclonal Antibody for IHC (P), Western Blot

This gene encodes glucose-6-phosphate dehydrogenase. This protein is a cytosolic enzyme encoded by a housekeeping X-linked gene whose main function is to produce NADPH, a key electron donor in the defense against oxidizing agents and in reductive biosynthetic reactions. G6PD is remarkable for its genetic diversity. Many variants of G6PD, mostly produced from missense mutations, have been described with wide ranging levels of enzyme activity and associated clinical symptoms. G6PD deficiency may cause neonatal jaundice, acute hemolysis, or severe chronic non-spherocytic hemolytic anemia. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G6PD
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1R70
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene G6PD
Gene Accession No. P05370, P11413
Gene Alias G28A; G6PD; G6PD1; G6pd-1; G6pdx; Glucose-6-phosphate 1-dehydrogenase; glucose-6-phosphate 1-dehydrogenase X; glucose-6-phosphate dehydrogenase; glucose-6-phosphate dehydrogenase X-linked; Gpdx
Gene Symbols G6PD
Host Species Rabbit
Immunogen Recombinant protein of human Glucose 6 Phosphate Dehydrogenase.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24377, 2539
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.