missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ G6PD Recombinant Rabbit Monoclonal Antibody (23GB1795), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | G6PD |
|---|---|
| Clone | 23GB1795 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088261
![]() |
Invitrogen™
MA554022 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087657
|
Invitrogen™
MA554023 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
G6PD Recombinant Monoclonal antibody specifically detects G6PD in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| G6PD | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P11413 | |
| 2539 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1795 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| G28A; G6PD; G6PD1; G6pd-1; G6pdx; Glucose-6-phosphate 1-dehydrogenase; glucose-6-phosphate 1-dehydrogenase X; glucose-6-phosphate dehydrogenase; glucose-6-phosphate dehydrogenase X-linked; Gpdx | |
| A synthesized peptide derived from human G6PD (460-515AA). | |
| Primary | |
| -20°C | |
| G6PD |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title