Learn More
G6PD Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25413-1-AP-20UL
 View more versions of this product
Product Code. 16883985
  • £135.00 / 20µL
Description
This gene encodes glucose-6-phosphate dehydrogenase. This protein is a cytosolic enzyme encoded by a housekeeping X-linked gene whose main function is to produce NADPH, a key electron donor in the defense against oxidizing agents and in reductive biosynthetic reactions. G6PD is remarkable for its genetic diversity. Many variants of G6PD, mostly produced from missense mutations, have been described with wide ranging levels of enzyme activity and associated clinical symptoms. G6PD deficiency may cause neonatal jaundice, acute hemolysis, or severe chronic non-spherocytic hemolytic anemia. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
G6PD
Polyclonal
Unconjugated
G6PD
G6PD, G6PD1
Rabbit
Antigen Affinity Chromatography
RUO
2539
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P11413
G6PD
G6PD Fusion Protein Ag21862
20 μL