missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ G6PC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109749
This item is not returnable.
View return policy
Description
Immunogen sequence: TLGAGIVIAE ALQNQLAWLE NVWLWITFLG DPKILFLFYF PAAYYASRRV GIAVLWISLI TEWLNLIFKW FLFGDRPFWW VHESGYYSQA PAQVHQFPSS.
Hydrolyzes glucose-6-phosphate to glucose in the endoplasmic reticulum. May form with the glucose-6-phosphate transporter (SLC37A4/G6PT) a ubiquitously expressed complex responsible for glucose production through glycogenolysis and gluconeogenesis. Probably required for normal neutrophil function.
Specifications
| G6PC3 | |
| Polyclonal | |
| Unconjugated | |
| G6PC3 | |
| 0710001K01Rik; 3.1.3.9; AU019276; AU045429; AV128920; AW545836; G6Pase 3; G-6-Pase 3; G6Pase-beta; G6pc3; glucose 6 phosphatase catalytic subunit 3; glucose 6 phosphatase, catalytic, 3; glucose-6-phosphatase 3; Glucose-6-phosphatase beta; glucose-6-phosphatase catalytic subunit 3; glucose-6-phosphatase catalytic subunit-related; glucose-6-phosphatase catalytic subunit-related protein; SCN4; Ubiquitous glucose-6-phosphatase catalytic subunit-related protein; ubiquitously expressed G6Pase catalytic subunit-related protein; UGRP; zgc:158425 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 303565, 68401, 92579 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.36 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q6AZ83, Q6NSQ9, Q9BUM1 | |
| G6PC3 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction