missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ G6PC Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals™ NBP3-27858-100ul

 View more versions of this product

Product Code. 30041624

  • £290.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

G6PC Polyclonal antibody specifically detects G6PC in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

G6PC
Polyclonal
Western Blot 1:500-2000, Immunohistochemistry 1:50-200
EC 3.1.3.9, G6Pase, G-6-Pase, G6Pase-alpha, G6PT, glucose-6-phosphatase, Glucose-6-phosphatase alpha, glucose-6-phosphatase, catalytic (glycogen storage disease type I, von Gierkedisease), glucose-6-phosphatase, catalytic subunit, GSD1, GSD1a, MGC163350
Synthesized peptide derived from human C-terminal G6PC (Uniprot #: P35575)
100 μL
Primary
Human, Mouse, Rat
Purified
Immunohistochemistry, Western Blot
Unconjugated
PBS, 50% glycerol
Rabbit
Antigen Affinity-purified
RUO
2538
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.