missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ G6PC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-27858-50ul
This item is not returnable.
View return policy
Description
G6PC Polyclonal antibody specifically detects G6PC in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| G6PC | |
| Polyclonal | |
| Western Blot 1:500-2000, Immunohistochemistry 1:50-200 | |
| EC 3.1.3.9, G6Pase, G-6-Pase, G6Pase-alpha, G6PT, glucose-6-phosphatase, Glucose-6-phosphatase alpha, glucose-6-phosphatase, catalytic (glycogen storage disease type I, von Gierkedisease), glucose-6-phosphatase, catalytic subunit, GSD1, GSD1a, MGC163350 | |
| Synthesized peptide derived from human C-terminal G6PC (Uniprot #: P35575) | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| PBS, 50% glycerol | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 2538 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction