Learn More
G6PC Antibody (CL5818) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61433G
Product Code. 30304804
  • £558.00 / 0.10mL
Description
G6PC Monoclonal antibody specifically detects G6PC in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
G6PC
Monoclonal
DyLight 488
EC 3.1.3.9, G6Pase, G-6-Pase, G6Pase-alpha, G6PT, glucose-6-phosphatase, Glucose-6-phosphatase alpha, glucose-6-phosphatase, catalytic (glycogen storage disease type I, von Gierkedisease), glucose-6-phosphatase, catalytic subunit, GSD1, GSD1a, MGC163350
This antibody was developed using a synthetic peptide derived from P35575, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL5818
50mM Sodium Borate
Mouse
Protein A purified
RUO
2538
Store at 4⪚C in the dark.
IgG2b