missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
G6PC Antibody (CL5817) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61432AF750
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
G6PC Monoclonal antibody specifically detects G6PC in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Tekniske data
| G6PC | |
| Monoclonal | |
| Alexa Fluor 750 | |
| EC 3.1.3.9, G6Pase, G-6-Pase, G6Pase-alpha, G6PT, glucose-6-phosphatase, Glucose-6-phosphatase alpha, glucose-6-phosphatase, catalytic (glycogen storage disease type I, von Gierkedisease), glucose-6-phosphatase, catalytic subunit, GSD1, GSD1a, MGC163350 | |
| This antibody was developed using a synthetic peptide derived from P35575, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL5817 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 2538 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion