missing translation for 'onlineSavingsMsg'
Learn More

G5pr Antibody, Novus Biologicals™

Product Code. 18336330 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336330 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336330 Supplier Novus Biologicals Supplier No. NBP287480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

G5pr Polyclonal specifically detects G5pr in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G5pr
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias C14orf10, chromosome 14 open reading frame 10, FLJ20644, G4-1, G5pr, protein phosphatase 2 (formerly 2A), regulatory subunit B′′, gamma, protein phosphatase 2, regulatory subunit B′′, gamma, Protein phosphatase subunit G5PR, rhabdomyosarcoma antigen MU-RMS-40.6A, Rhabdomyosarcoma antigen MU-RMS-40.6A/6C, rhabdomyosarcoma antigen Mu-RMS-40.6C, serine/threonine-protein phosphatase 2A regulatory subunit B′′ subunit gamma
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of human G5pr
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55012
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.