missing translation for 'onlineSavingsMsg'
Learn More

Tocris™ G5
GREENER_CHOICE

Product Code. p-7166820
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18745786

Brand: Tocris™ 6776/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ubiquitin isopeptidase inhibitor; induces apoptosis

G5 is an ubiquitin isopeptidase inhibitor; activates caspases and apoptosis in an apoptosome-independent pathway.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Molecular Formula C19H14N2O7S
Formula Weight 414.39
Product Type Inhibitor
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.