missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ G5

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
G5 is an ubiquitin isopeptidase inhibitor; activates caspases and apoptosis in an apoptosome-independent pathway.
Specifications
Specifications
| Quantity | 10 mg |
| Molecular Formula | C19H14N2O7S |
| Formula Weight | 414.39 |
| Product Type | Inhibitor |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction